Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX648474.1 - 1 2649 Chr. 13 - 52014528 - 52017668 [-] on Build GRCh38 98


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ALG11, alpha-1,2-mannosyltransferase ALG11 CDG1P, GT8 Anti-sense Chr. - 52012387 - 52029648 on Build GRCh38
Downstream UTP14, small subunit processome component homolog C (S. cerevisiae) UTP14C 2700066J21Rik, KIAA0266, UTP14B 7.0kb Anti-sense Chr. - 52024691 - 52033600 on Build GRCh38

View Full Product Details