Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CIITA
- Entrez Gene ID
-
4261
- Gene Name
- class II, major histocompatibility complex, transactivator
- Gene Aliases
-
C2TA, CIITAIV, MHC2TA, NLRA
- Chromosome Location
-
Chr.16: 10866208 - 10941562 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CB956280.1 |
- |
1 |
293 |
|
Chr. 16 - 10935165 - 10935624 [+] on Build GRCh38 |
92 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
trans-golgi network vesicle protein 23 homolog A |
TVP23A |
FAM18A, YDR084C |
-115.0kb
|
Anti-sense |
Chr. - 10758034 - 10818902 on Build GRCh38 |
Downstream |
C-type lectin domain family 16 member A |
CLEC16A |
Gop-1, KIAA0350 |
8.2kb
|
Sense |
Chr. - 10944488 - 11193272 on Build GRCh38 |
View Full Product Details