Assay Design

The assay probe spans an exon junction.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK130715.1 - 1 - 2 206 Chr. 1 - 115283034 - 115368263 [+] on Build GRCh38 67


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream tetraspanin 2 TSPAN2 NET3, TSN2, TSPAN-2 -193.5kb Anti-sense Chr. - 115048011 - 115089501 on Build GRCh38
Downstream VANGL planar cell polarity protein 1 VANGL1 KITENIN, LPP2, STB2, STBM2 273.7kb Sense Chr. - 115641953 - 115698224 on Build GRCh38

View Full Product Details