Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK130779.1 - 1 447 Chr. 3 - 113004921 - 113006674 [-] on Build GRCh38 71


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream GTP binding protein 8 (putative) GTPBP8 HSPC135 -3.5kb Anti-sense Chr. - 112989275 - 113001966 on Build GRCh38
Downstream uncharacterized LOC101929717 LOC101929717 135.7kb Sense Chr. - 113142350 - 113167731 on Build GRCh38

View Full Product Details