The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252039.1 | NP_001238968.1 | 4 - 5 | 771 | 57 |
NM_004583.3 | NP_004574.2 | 3 - 4 | 555 | 57 | |
NM_201434.2 | NP_958842.1 | 4 - 5 | 672 | 57 | |
XM_011525090.1 | XP_011523392.1 | 3 - 4 | 494 | 57 | |
XM_011525091.1 | XP_011523393.1 | 3 - 4 | 618 | 57 | |
GenBank mRNA | AF141304.1 | - | 2 - 3 | 384 | 57 |
AF498938.1 | - | 2 - 3 | 315 | 57 | |
AK310234.1 | - | 4 - 5 | 715 | 57 | |
AL553867.3 | - | 3 - 4 | 510 | 57 | |
BC106039.1 | - | 3 - 4 | 514 | 57 | |
BC114439.1 | - | 3 - 4 | 510 | 57 | |
BM424166.1 | - | 4 - 5 | 612 | 57 | |
BM550607.1 | - | 3 - 4 | 515 | 57 | |
BT019484.1 | - | 2 - 3 | 315 | 57 | |
CR541901.1 | - | 2 - 3 | 315 | 57 | |
U11293.1 | - | 2 - 3 | 390 | 57 | |
U18420.1 | - | 3 - 4 | 450 | 57 |