Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK055756.1 - 1 1251 Chr. 10 - 62074903 - 62076561 [+] on Build GRCh38 81
GenBankmRNA DB320410.1 - 1 Chr. 10 - 62076025 - 62076561 [-] on Build GRCh38 81


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream chromosome 10 open reading frame 107 C10orf107 bA63A2.1 -308.1kb Sense Chr. - 61662927 - 61766766 on Build GRCh38
Downstream rhotekin 2 RTKN2 PLEKHK1, bA531F24.1 106.5kb Anti-sense Chr. - 62159131 - 62268879 on Build GRCh38

View Full Product Details