Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK055821.1 - 1 - 2 388 Chr. 2 - 235727565 - 235730477 [+] on Build GRCh38 91
GenBankmRNA DA625705.1 - 1 - 2 388 Chr. 2 - 235727565 - 235728296 [+] on Build GRCh38 91


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream AGAP1 intronic transcript 1 AGAP1-IT1 -220.0kb Sense Chr. - 235505751 - 235507566 on Build GRCh38
Downstream uncharacterized LOC105373942 LOC105373942 15.9kb Anti-sense Chr. - 235746235 - 235791606 on Build GRCh38

View Full Product Details