Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PRPF18
- Entrez Gene ID
-
8559
- Gene Name
- pre-mRNA processing factor 18
- Gene Aliases
-
PRP18, hPrp18
- Chromosome Location
-
Chr.10: 13586811 - 13645279 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK056870.1 |
- |
1 - 1 |
2551 |
104 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB329724.1 |
- |
1 |
|
|
Chr. 10 - 13635683 - 13636258 [-] on Build GRCh38 |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101928524 |
LOC101928524 |
|
|
Sense |
Chr. - 13650927 - 13655929 on Build GRCh38 |
Downstream |
FERM domain containing 4A |
FRMD4A |
CCAFCA, FRMD4, bA295P9.4 |
7.4kb
|
Anti-sense |
Chr. - 13643706 - 14330952 on Build GRCh38 |
View Full Product Details