Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK057219.1 - 1 1156 Chr. X - 24488887 - 24491278 [+] on Build GRCh38 76


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream SPT20 homolog, SAGA complex component-like 1 SUPT20HL1 FAM48B1, SPT20L -123.5kb Sense Chr. - 24362761 - 24365424 on Build GRCh38
Downstream phosphate cytidylyltransferase 1, choline, beta PCYT1B CCTB, CTB 66.8kb Anti-sense Chr. - 24558087 - 24672862 on Build GRCh38

View Full Product Details