Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FANCA
- Entrez Gene ID
-
2175
- Gene Name
- Fanconi anemia complementation group A
- Gene Aliases
-
FA, FA-H, FA1, FAA, FACA, FAH, FANCH
- Chromosome Location
-
Chr.16: 89737551 - 89816658 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY339879.1 |
- |
1 - 1 |
146 |
107 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
zinc finger protein 276 |
ZNF276 |
CENP-Z, CENPZ, ZADT, ZFP276, ZNF477 |
-41.9kb
|
Anti-sense |
Chr. - 89720368 - 89740925 on Build GRCh38 |
Downstream |
spire type actin nucleation factor 2 |
SPIRE2 |
Spir-2 |
34.4kb
|
Anti-sense |
Chr. - 89817477 - 89871319 on Build GRCh38 |
View Full Product Details