Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FGB
- Entrez Gene ID
-
2244
- Gene Name
- fibrinogen beta chain
- Gene Aliases
-
HEL-S-78p
- Chromosome Location
-
Chr.4: 154562980 - 154572763 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC060040.1 |
- |
1 - 1 |
1654 |
71 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
pleiotropic regulator 1 |
PLRG1 |
Cwc1, PRL1, PRP46, PRPF46, TANGO4 |
-12.6kb
|
Anti-sense |
Chr. - 154534997 - 154550433 on Build GRCh38 |
Downstream |
fibrinogen alpha chain |
FGA |
Fib2 |
18.4kb
|
Anti-sense |
Chr. - 154583126 - 154590766 on Build GRCh38 |
View Full Product Details