Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- COMMD5
- Entrez Gene ID
-
28991
- Gene Name
- COMM domain containing 5
- Gene Aliases
-
HCARG, HT002
- Chromosome Location
-
Chr.8: 144837978 - 144853578 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK092477.1 |
- |
1 - 1 |
449 |
92 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA843501.1 |
- |
1 |
449 |
|
Chr. 8 - 144839360 - 144839944 [-] on Build GRCh38 |
92 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
zinc finger protein 517 |
ZNF517 |
|
-29.7kb
|
Anti-sense |
Chr. - 144798876 - 144813033 on Build GRCh38 |
Downstream |
zinc finger protein 250 |
ZNF250 |
ZFP647, ZNF647 |
37.0kb
|
Sense |
Chr. - 144876951 - 144902311 on Build GRCh38 |
View Full Product Details