Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- METTL22
- Entrez Gene ID
-
79091
- Gene Name
- methyltransferase like 22
- Gene Aliases
-
C16orf68
- Chromosome Location
-
Chr.16: 8621640 - 8673565 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK097189.1 |
- |
1 |
1548 |
|
Chr. 16 - 8650168 - 8652458 [+] on Build GRCh38 |
107 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
transmembrane protein 114 |
TMEM114 |
|
-60.0kb
|
Anti-sense |
Chr. - 8526549 - 8590193 on Build GRCh38 |
Downstream |
4-aminobutyrate aminotransferase |
ABAT |
GABA-AT, GABAT, NPD009 |
22.1kb
|
Sense |
Chr. - 8674587 - 8784575 on Build GRCh38 |
View Full Product Details