Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
Z39243.1 |
- |
|
|
|
Chr. - - [-] on Build |
77 |
GenBankmRNA |
BF921508.1 |
- |
1 |
|
|
Chr. 15 - 89509812 - 89510388 [-] on Build GRCh38 |
77 |
GenBankmRNA |
AA453981.1 |
- |
1 |
|
|
Chr. 15 - 89509973 - 89510325 [+] on Build GRCh38 |
78 |
GenBankmRNA |
AL109703.1 |
- |
1 |
1350 |
|
Chr. 15 - 89508961 - 89510416 [+] on Build GRCh38 |
77 |
GenBankmRNA |
AI816364.1 |
- |
1 |
|
|
Chr. 15 - 89509913 - 89510416 [-] on Build GRCh38 |
77 |
GenBankmRNA |
AA453473.1 |
- |
1 |
|
|
Chr. 15 - 89510048 - 89510416 [-] on Build GRCh38 |
77 |
GenBankmRNA |
AA676759.1 |
- |
1 |
|
|
Chr. 15 - 89510083 - 89510414 [-] on Build GRCh38 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 928 |
LINC00928 |
|
|
Anti-sense |
Chr. - 89504930 - 89524034 on Build GRCh38 |
Downstream |
TOPBP1 interacting checkpoint and replication regulator |
TICRR |
C15orf42, SLD3, Treslin |
65.0kb
|
Sense |
Chr. - 89575587 - 89628023 on Build GRCh38 |
View Full Product Details