The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_046762.1 | 1 - 1 | 52 | 98 | |
GenBank mRNA | BC041487.1 | - | 1 - 1 | 52 | 98 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | ALMS1, centrosome and basal body associated protein | ALMS1 | ALSS | Sense | Chr. - 73385758 - 73609919 on Build GRCh38 | |
Downstream | N-acetyltransferase 8 (putative) | NAT8 | ATase2, CCNAT, CML1, GLA, Hcml1, TSC501, TSC510 | 183.4kb | Anti-sense | Chr. - 73640723 - 73642410 on Build GRCh38 |