Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
APRT | NM_000485.2 | NP_000476.1 | ||
NM_001030018.1 | NP_001025189.1 | |||
BC107151.2 | ||||
BM550173.1 | ||||
CR749423.1 | CAH18261.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv37n68 | Chr16:88767460 - 88944903 on Build GRCh38 | Loss | LOC101927793 CDT1 LOC105371409 PABPN1L APRT LOC100129697 PIEZO1 LOC107987238 GALNS TRAPPC2L CBFA2T3 |
nsv1063024 | Chr16:88707330 - 89238482 on Build GRCh38 | Gain | LOC100289580 PABPN1L APRT ZNF778 MIR4722 LOC339059 SLC22A31 LOC107987238 CTU2 LINC00304 LOC101927793 CDT1 LOC105371409 LOC100129697 PIEZO1 GALNS ACSF3 LOC400558 TRAPPC2L CDH15 CBFA2T3 |
nsv471115 | Chr16:88801844 - 89197442 on Build GRCh38 | Loss | PABPN1L APRT SLC22A31 LOC107987238 LINC00304 LOC101927793 CDT1 LOC105371409 LOC100129697 GALNS ACSF3 LOC400558 TRAPPC2L CDH15 CBFA2T3 |
dgv3064n100 | Chr16:88683753 - 88846491 on Build GRCh38 | Gain | CDT1 SNAI3-AS1 RNF166 LOC100289580 APRT MIR4722 LOC339059 PIEZO1 LOC107987238 CTU2 GALNS SNAI3 |
nsv573718 | Chr16:88807259 - 88810716 on Build GRCh38 | Loss | CDT1 APRT |
nsv573674 | Chr16:88738775 - 89325907 on Build GRCh38 | Gain | LOC100289580 PABPN1L APRT ZNF778 LOC100287036 LOC339059 SLC22A31 LOC107987238 LINC00304 LOC101927793 CDT1 LOC105371409 LOC100129697 PIEZO1 ANKRD11 GALNS ACSF3 LOC400558 TRAPPC2L LOC105371414 CDH15 CBFA2T3 |
esv3639555 | Chr16:88745961 - 88899576 on Build GRCh38 | Gain | LOC101927793 CDT1 PABPN1L APRT PIEZO1 LOC107987238 GALNS TRAPPC2L CBFA2T3 |
nsv471117 | Chr16:88805821 - 88838392 on Build GRCh38 | Gain | CDT1 APRT LOC107987238 GALNS |
nsv952071 | Chr16:88790893 - 89019992 on Build GRCh38 | Deletion | LOC101927793 CDT1 LOC105371409 PABPN1L APRT LOC100129697 LOC107987238 GALNS TRAPPC2L CBFA2T3 |
nsv482951 | Chr16:88633593 - 90228345 on Build GRCh38 | Loss | TCF25 LOC107987238 LOC107984817 SNORD68 RPL13 GAS8-AS1 CDK10 CENPBD1 URAHP IL17C AFG3L1P PABPN1L CYBA APRT ZNF778 MIR4722 LOC339059 CPNE7 SPIRE2 CTU2 FAM157C PRDM7 LOC105371409 LOC105371419 LOC100129697 PIEZO1 ACSF3 LOC400558 MVD TRAPPC2L LOC105371414 VPS9D1-AS1 SLC22A31 DBNDD1 GAS8 CDT1 LOC101927817 CHMP1A SPATA33 ANKRD11 ZNF276 TUBB3 CBFA2T3 LOC100289580 LOC100287036 SPATA2L VPS9D1 SPG7 TUBB8P7 LINC00304 LOC101927793 DPEP1 SNAI3-AS1 RNF166 MC1R FANCA DEF8 GALNS SNAI3 CDH15 |
nsv1063441 | Chr16:88767066 - 88861882 on Build GRCh38 | Loss | CDT1 APRT PIEZO1 LOC107987238 GALNS TRAPPC2L |
dgv3065n100 | Chr16:88711587 - 88927172 on Build GRCh38 | Gain | LOC100289580 PABPN1L APRT MIR4722 LOC339059 LOC107987238 CTU2 LOC101927793 CDT1 PIEZO1 GALNS TRAPPC2L CBFA2T3 |