Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CDKN2B
- Entrez Gene ID
-
1030
- Gene Name
- cyclin dependent kinase inhibitor 2B
- Gene Aliases
-
CDK4I, INK4B, MTS2, P15, TP15, p15INK4b
- Chromosome Location
-
Chr.9: 22002903 - 22009313 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF488411.1 |
- |
- |
451 |
95 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA134897.1 |
- |
1 |
72 |
|
Chr. 8 - 127204885 - 127205273 [-] on Build GRCh38 |
95 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cancer susceptibility candidate 19 (non-protein coding) |
CASC19 |
CARLo-6, LINC01245 |
-7.3kb
|
Sense |
Chr. - 127187785 - 127197627 on Build GRCh38 |
Downstream |
colon cancer associated transcript 1 (non-protein coding) |
CCAT1 |
CARLo-5, onco-lncRNA-40 |
2.1kb
|
Sense |
Chr. - 127207382 - 127219268 on Build GRCh38 |
View Full Product Details