Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AU117938.1 - 1 Chr. 15 - 64955258 - 64955892 [-] on Build GRCh38 92
GenBankmRNA DB129769.1 - 2 333 Chr. 15 - 64955361 - 64957230 [-] on Build GRCh38 92
GenBankmRNA AK021773.1 - 1 550 Chr. 15 - 64952058 - 64956157 [-] on Build GRCh38 92
GenBankmRNA AV763456.1 - 2 348 Chr. 15 - 64955194 - 64957251 [-] on Build GRCh38 92
GenBankmRNA AA846950.1 - 1 Chr. 15 - 64955427 - 64955781 [+] on Build GRCh38 92
GenBankmRNA AU119153.1 - 1 550 Chr. 15 - 64955334 - 64956157 [-] on Build GRCh38 92


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream pleckstrin homology domain containing O2 PLEKHO2 PLEKHQ1, PP1628, pp9099 -86.5kb Anti-sense Chr. - 64841883 - 64868002 on Build GRCh38
Downstream spastic paraplegia 21 (autosomal recessive, Mast syndrome) SPG21 ACP33, BM-019, GL010, MAST 6.9kb Sense Chr. - 64963021 - 64989946 on Build GRCh38

View Full Product Details