Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AJ006345.3 - 1 22435 Chr. 11 - 2444100 - 2849207 [+] on Build GRCh38 99
GenBankmRNA AK226077.1 - 1 4645 Chr. 11 - 2461889 - 2467088 [+] on Build GRCh38 99


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream transient receptor potential cation channel subfamily M member 5 TRPM5 LTRPC5, MTR1 -26.9kb Anti-sense Chr. - 2403962 - 2423424 on Build GRCh38
Downstream KCNQ1 opposite strand/antisense transcript 1 (non-protein coding) KCNQ1OT1 KCNQ1-AS2, KCNQ10T1, Kncq1, KvDMR1, KvLQT1-AS, LIT1, NCRNA00012 141.2kb Anti-sense Chr. - 2608328 - 2699998 on Build GRCh38

View Full Product Details