Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RPL6P28
- Entrez Gene ID
-
100271208
- Gene Name
- ribosomal protein L6 pseudogene 28
- Gene Aliases
-
RPL6_16_1757
- Chromosome Location
-
Chr.22: 44901571 - 44902450 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC037979.1 |
- |
1 - 1 |
50 |
75 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BI522604.1 |
- |
1 |
50 |
|
Chr. 22 - 44901547 - 44902458 [+] on Build GRCh38 |
75 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
PRR5-ARHGAP8 readthrough |
PRR5-ARHGAP8 |
|
-38.8kb
|
Sense |
Chr. - 44702198 - 44862784 on Build GRCh38 |
Downstream |
uncharacterized LOC101927551 |
LOC101927551 |
|
95.3kb
|
Anti-sense |
Chr. - 44997734 - 45003619 on Build GRCh38 |
View Full Product Details