Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOC642366
- Entrez Gene ID
-
642366
- Gene Name
- uncharacterized LOC642366
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 51372736 - 51383332 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_046243.1 |
|
2 - 2 |
5679 |
104 |
GenBank mRNA |
CR936786.1 |
- |
1 - 1 |
5743 |
105 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CK818674.1 |
- |
1 |
510 |
|
Chr. 5 - 51373678 - 51374265 [-] on Build GRCh38 |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC100287592 |
LOC100287592 |
|
-402.5kb
|
Sense |
Chr. - 50969217 - 50970187 on Build GRCh38 |
Downstream |
ISL LIM homeobox 1 |
ISL1 |
ISLET1, Isl-1 |
3.6kb
|
Anti-sense |
Chr. - 51383124 - 51394730 on Build GRCh38 |
View Full Product Details