Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FUCA1
Entrez Gene ID
2517
Gene Name
fucosidase, alpha-L- 1, tissue
Gene Aliases
FUCA
Chromosome Location
Chr.1: 23845077 - 23868369 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000147.4 NP_000138.2 6 - 7 1235 69
XM_005245821.2 XP_005245878.1 6 - 7 1372 69
XM_011541167.2 XP_011539469.1 6 - 7 820 69
XM_017000905.1 XP_016856394.1 6 - 7 917 69
GenBank mRNA AB074175.1 - 6 - 7 1156 69
AK092914.1 - 7 - 8 1090 69
AK095443.1 - 7 - 8 1107 69
AK225468.1 - 6 - 7 1156 69
AK301302.1 - 8 - 9 1085 69
AK314649.1 - 6 - 7 1156 69
BC017338.2 - 6 - 7 1158 69
M10355.1 - 6 - 7 930 69
M29877.1 - 6 - 7 1155 69
X01390.1 - 6 - 7 930 69

View Full Product Details