The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134774.1 | NP_001128246.1 | 2 - 3 | 406 | 84 |
GenBank mRNA | AK094593.1 | - | 1 - 2 | 276 | 84 |
BE882786.1 | - | 2 - 3 | 301 | 84 |