Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SKA3
Entrez Gene ID
221150
Gene Name
spindle and kinetochore associated complex subunit 3
Gene Aliases
C13orf3, RAMA1
Chromosome Location
Chr.13: 21153595 - 21176602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166017.1 NP_001159489.1 1 - 2 231 79
NM_145061.5 NP_659498.4 1 - 2 231 79
GenBank mRNA AF361358.1 - 1 - 2 173 79
AK090584.1 - 1 - 2 176 79
BC048988.1 - 1 - 2 202 79
BI460315.1 - 1 - 2 206 79

View Full Product Details