Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SKA3
Entrez Gene ID
221150
Gene Name
spindle and kinetochore associated complex subunit 3
Gene Aliases
C13orf3, RAMA1
Chromosome Location
Chr.13: 21153595 - 21176602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166017.1 NP_001159489.1 4 - 5 871 196
NM_145061.5 NP_659498.4 4 - 5 871 196
XM_005266288.3 XP_005266345.1 3 - 4 657 196
XM_011534994.1 XP_011533296.1 4 - 5 847 196
GenBank mRNA AF361358.1 - 4 - 5 813 196
AK090584.1 - 4 - 5 816 196
AK307976.1 - 5 - 6 746 196
AK314864.1 - 4 - 5 817 196
BC013418.2 - 3 - 4 755 196
BC048988.1 - 4 - 5 842 196

View Full Product Details