Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
FOXP3 | NM_001114377.1 | NP_001107849.1 | ||
NM_014009.3 | NP_054728.2 | |||
XM_006724533.2 | XP_006724596.2 | |||
XM_011543916.2 | XP_011542218.1 | |||
XM_017029565.1 | XP_016885054.1 | |||
XM_017029566.1 | XP_016885055.1 | |||
XM_017029567.1 | XP_016885056.1 | |||
AF277993.1 | AAG53607.1 | |||
AJ005891.1 | CAA06748.1 | |||
AK292052.1 | ||||
AK299988.1 | ||||
BC113401.1 | ||||
BC113403.1 | ||||
BC143785.1 | ||||
BC143786.1 | ||||
DQ010327.1 | AAY27088.1 | |||
EF534714.1 | ||||
EU855812.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471602 | ChrX:49205142 - 49320261 on Build GRCh38 | Gain | FOXP3 CCDC22 PPP1R3F CACNA1F GAGE10 |
nsv528292 | ChrX:48433286 - 49274368 on Build GRCh38 | Gain | SLC35A2 FOXP3 GPKOW PPP1R3F WAS RBM3 TFE3 MAGIX SYP GATA1 LOC101927635 FTSJ1 PIM2 WDR45 PRICKLE3 PRAF2 PCSK1N TIMM17B SLC38A5 WDR13 PQBP1 SYP-AS1 GLOD5 TBC1D25 GRIPAP1 HDAC6 PORCN CACNA1F EBP PLP2 CCDC22 KCND1 CCDC120 ERAS SUV39H1 OTUD5 |
esv32909 | ChrX:48003374 - 49285646 on Build GRCh38 | Gain+Loss | SLC35A2 FOXP3 GPKOW PPP1R3F ZNF630 SSX1 WAS RBM3 SSX5 SPACA5 SSX6 TFE3 MAGIX SYP SSX4 GATA1 LOC101927635 FTSJ1 PIM2 WDR45 PRICKLE3 PRAF2 PCSK1N SSX4B TIMM17B SLC38A5 WDR13 PQBP1 SYP-AS1 GLOD5 TBC1D25 ZNF182 GRIPAP1 HDAC6 PORCN CACNA1F ZNF630-AS1 EBP PLP2 SSX3 CCDC22 KCND1 SPACA5B CCDC120 SSX9 ERAS SUV39H1 OTUD5 |
nsv469734 | ChrX:49205141 - 49320261 on Build GRCh38 | Gain+Loss | FOXP3 CCDC22 PPP1R3F CACNA1F GAGE10 |
nsv6895 | ChrX:49227319 - 49262690 on Build GRCh38 | Insertion | FOXP3 CCDC22 CACNA1F |