Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFYVE28
Entrez Gene ID
57732
Gene Name
zinc finger FYVE-type containing 28
Gene Aliases
LST2, LYST2
Chromosome Location
Chr.4: 2269585 - 2418698 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172656.1 NP_001166127.1 2 - 3 499 74
NM_001172657.1 NP_001166128.1 2 - 3 499 74
NM_001172659.1 NP_001166130.1 2 - 3 532 74
NM_001172660.1 NP_001166131.1 2 - 3 532 74
NM_020972.2 NP_066023.2 2 - 3 499 74
XM_006713900.3 XP_006713963.1 2 - 3 553 74
XM_006713902.3 XP_006713965.1 2 - 3 554 74
XM_006713904.3 XP_006713967.1 2 - 3 553 74
XM_011513541.2 XP_011511843.1 2 - 3 553 74
XM_017008502.1 XP_016863991.1 2 - 3 275 74
XM_017008503.1 XP_016863992.1 2 - 3 554 74
XM_017008504.1 XP_016863993.1 2 - 3 553 74
XM_017008505.1 XP_016863994.1 2 - 3 275 74
XM_017008506.1 XP_016863995.1 2 - 3 554 74
XM_017008507.1 XP_016863996.1 2 - 3 553 74
XM_017008508.1 XP_016863997.1 2 - 3 553 74
XM_017008509.1 XP_016863998.1 2 - 3 553 74
XM_017008510.1 XP_016863999.1 2 - 3 553 74
GenBank mRNA AB046863.1 - 2 - 3 498 74
AK294710.1 - 3 - 4 326 74
AK316484.1 - 2 - 3 532 74
BC053664.1 - 2 - 3 465 74
BC137309.1 - 3 - 4 242 74
BC137310.1 - 3 - 4 242 74

View Full Product Details