The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_054023.4 | NP_473364.1 | 2 - 3 | 350 | 75 |
GenBank mRNA | AA988559.1 | - | 1 - 2 | 75 | |
AF313455.1 | - | 2 - 3 | 271 | 75 | |
AF439545.1 | - | 2 - 3 | 339 | 75 | |
AY358979.1 | - | 2 - 3 | 339 | 75 | |
BC024232.1 | - | 2 - 3 | 354 | 75 |