Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NCOA5
Entrez Gene ID
57727
Gene Name
nuclear receptor coactivator 5
Gene Aliases
CIA, bA465L10.6
Chromosome Location
Chr.20: 46060985 - 46089962 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020967.2 NP_066018.1 4 - 4 592 65
XM_005260474.3 XP_005260531.1 3 - 3 546 65
XM_011528951.2 XP_011527253.1 4 - 4 607 65
XM_017027988.1 XP_016883477.1 3 - 3 546 65
GenBank mRNA AB046857.1 - 2 - 2 140 65
AF230533.1 - 4 - 4 564 65
AF470686.1 - 3 - 3 968 65
BC140836.1 - 3 - 3 481 65
BC151133.1 - 3 - 3 481 65

View Full Product Details