The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012989.2 | NP_001013007.1 | 1 - 1 | 48 | 67 |
NR_121210.1 | 1 - 1 | 48 | 67 | ||
GenBank mRNA | GU727624.1 | - | 1 - 1 | 48 | 67 |