The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001015879.1 | NP_001015879.1 | 1 - 2 | 105 | 59 |
GenBank mRNA | AY661554.1 | - | 1 - 2 | 59 |