Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM91
Entrez Gene ID
641649
Gene Name
transmembrane protein 91
Gene Aliases
DSPC3, IFITMD6
Chromosome Location
Chr.19: 41363966 - 41384083 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098822.1 NP_001092292.1 3 - 4 715 74
NM_001098825.1 NP_001092295.1 3 - 4 523 74
GenBank mRNA AI214374.1 - 1 - 2 74
BM726200.1 - 3 - 4 490 74
BX090160.1 - 2 - 3 398 74

View Full Product Details