The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198586.2 | NP_940988.2 | 1 - 1 | 1052 | 84 |
GenBank mRNA | AY324850.1 | - | 1 - 1 | 1052 | 84 |
BC103888.1 | - | 1 - 1 | 1052 | 84 | |
BC103889.1 | - | 1 - 1 | 1052 | 84 | |
BC103890.1 | - | 1 - 1 | 1052 | 84 |