The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198682.2 | NP_941391.2 | 3 - 4 | 307 | 102 |
XM_017008139.1 | XP_016863628.1 | 4 - 5 | 4234 | 102 | |
GenBank mRNA | BC017864.1 | - | 3 - 4 | 303 | 102 |
BC064396.1 | - | 3 - 4 | 264 | 102 | |
BG528203.1 | - | 3 - 4 | 303 | 102 |