The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199259.3 | NP_954868.1 | 4 - 5 | 408 | 71 |
NM_199260.3 | NP_954869.1 | 4 - 5 | 408 | 71 | |
NM_199261.3 | NP_954870.2 | 4 - 5 | 408 | 71 | |
GenBank mRNA | AF007118.1 | - | 4 - 5 | 356 | 71 |
AF495908.1 | - | 4 - 5 | 356 | 71 | |
AK308254.1 | - | 4 - 5 | 304 | 71 | |
AK314285.1 | - | 4 - 5 | 307 | 71 | |
AL833345.1 | - | 4 - 5 | 369 | 71 | |
AY219886.1 | - | 4 - 5 | 356 | 71 | |
AY219887.1 | - | 4 - 5 | 356 | 71 | |
BC028719.1 | - | 4 - 5 | 391 | 71 | |
DB459772.1 | - | 4 - 5 | 386 | 71 |