The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_203288.1 | NP_976033.1 | 2 - 3 | 200 | 78 |
GenBank mRNA | BC025928.1 | - | 2 - 3 | 200 | 78 |