Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCFD2
Entrez Gene ID
90411
Gene Name
multiple coagulation factor deficiency 2
Gene Aliases
F5F8D, F5F8D2, LMAN1IP, SDNSF
Chromosome Location
Chr.2: 46901870 - 46941855 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171511.2 NP_001164982.1 1 - 2 288 68
GenBank mRNA AK293889.1 - 1 - 2 172 68
DC321899.1 - 1 - 2 276 68
DC332658.1 - 1 - 2 288 68

View Full Product Details