The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002792.3 | NP_002783.1 | 3 - 4 | 504 | 93 |
GenBank mRNA | AF022815.1 | - | 3 - 4 | 373 | 93 |
AF054185.1 | - | 3 - 4 | 434 | 93 | |
AK312025.1 | - | 3 - 4 | 399 | 93 | |
BC001895.1 | - | 3 - 4 | 437 | 93 | |
BC004427.1 | - | 3 - 4 | 422 | 93 | |
BC014999.1 | - | 3 - 4 | 432 | 93 | |
BG575009.1 | - | 3 - 4 | 492 | 94 | |
BI258951.1 | - | 3 - 4 | 504 | 94 | |
BT007165.1 | - | 3 - 4 | 349 | 93 | |
BU632089.1 | - | 4 - 5 | 93 | ||
GQ472224.1 | - | 3 - 4 | 422 | 93 |