Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BNIP3
Entrez Gene ID
664
Gene Name
BCL2 interacting protein 3
Gene Aliases
NIP3
Chromosome Location
Chr.10: 131967683 - 131982013 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004052.3 NP_004043.3 1 - 2 257 92
GenBank mRNA AF002697.1 - 1 - 2 176 92
AK222626.1 - 1 - 2 167 92
AK295136.1 - 1 - 2 167 92
BC009342.2 - 1 - 2 152 92
BC021989.1 - 1 - 2 171 92
BC067818.1 - 1 - 2 200 92
BC080643.1 - 1 - 2 152 92
HY049371.1 - 1 - 2 258 92
U15174.1 - 1 - 2 121 92

View Full Product Details