The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001302123.1 | NP_001289052.1 | 1 - 2 | 268 | 96 |
NM_005409.4 | NP_005400.1 | 1 - 2 | 268 | 96 | |
GenBank mRNA | AF002985.1 | - | 1 - 2 | 145 | 96 |
AF030514.1 | - | 1 - 2 | 127 | 96 | |
AF352781.1 | - | 1 - 2 | 136 | 96 | |
AL703031.1 | - | 1 - 2 | 108 | 96 | |
BC005292.1 | - | 1 - 2 | 118 | 96 | |
BC012532.1 | - | 1 - 2 | 272 | 96 | |
BC110986.1 | - | 1 - 2 | 127 | 96 | |
BP289021.1 | - | 1 - 2 | 170 | 96 | |
BT006787.1 | - | 1 - 2 | 58 | 96 | |
U59286.1 | - | 1 - 2 | 80 | 96 | |
U66096.1 | - | 1 - 2 | 103 | 96 | |
Y15220.1 | - | 1 - 2 | 64 | 96 |