The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014501.2 | NP_055316.2 | 1 - 2 | 373 | 141 |
XM_011526752.1 | XP_011525054.1 | 1 - 2 | 562 | 141 | |
GenBank mRNA | AB062397.1 | - | 1 - 1 | 183 | 141 |
BC004236.2 | - | 1 - 1 | 183 | 141 | |
BC007554.1 | - | 1 - 1 | 183 | 141 | |
BC065364.1 | - | 1 - 1 | 193 | 141 | |
CN305224.1 | - | 1 - 2 | 374 | 142 |