Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTXN2
Entrez Gene ID
399697
Gene Name
cortexin 2
Gene Aliases
-
Chromosome Location
Chr.15: 48178564 - 48203754 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145668.1 NP_001139140.1 1 - 2 183 96
XM_017022178.1 XP_016877667.1 2 - 3 195 96
GenBank mRNA BX537772.1 - 1 - 2 183 96

View Full Product Details