The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016545.4 | NP_057629.2 | 1 - 1 | 152 | 64 |
GenBank mRNA | AI696140.1 | - | 1 - 1 | 152 | 64 |