The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152361.2 | NP_689574.1 | 1 - 1 | 460 | 74 |
GenBank mRNA | AK096263.1 | - | 1 - 1 | 447 | 74 |
BC009725.1 | - | 1 - 1 | 448 | 74 | |
BC040157.1 | - | 1 - 1 | 404 | 74 | |
BC051333.1 | - | 1 - 1 | 421 | 74 | |
DA382981.1 | - | 1 - 1 | 461 | 74 |