The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145356.1 | NP_001138828.1 | 1 - 2 | 258 | 101 |
NM_018491.3 | NP_060961.3 | 1 - 2 | 258 | 101 | |
XM_005251513.4 | XP_005251570.1 | 1 - 2 | 266 | 101 | |
XM_005251514.1 | XP_005251571.1 | 1 - 2 | 258 | 101 | |
XM_005251515.1 | XP_005251572.1 | 1 - 2 | 258 | 101 | |
XM_005251517.4 | XP_005251574.1 | 1 - 2 | 258 | 101 | |
XM_011517962.2 | XP_011516264.1 | 1 - 2 | 258 | 101 | |
XM_011517964.2 | XP_011516266.1 | 1 - 2 | 258 | 101 | |
XM_017014925.1 | XP_016870414.1 | 1 - 2 | 258 | 101 | |
GenBank mRNA | AF212253.1 | - | 1 - 2 | 226 | 101 |
AF257330.2 | - | 1 - 2 | 216 | 101 | |
AK315855.1 | - | 1 - 2 | 182 | 101 | |
AY320414.1 | - | 1 - 2 | 228 | 101 | |
AY343911.1 | - | 1 - 2 | 215 | 101 | |
BC005996.1 | - | 1 - 2 | 258 | 101 | |
BC013432.1 | - | 1 - 2 | 229 | 101 | |
BC132759.1 | - | 1 - 2 | 192 | 101 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172003.3 | NP_742000.1 | 1 - 2 | 330 | 101 |
XM_005263600.2 | XP_005263657.1 | 1 - 2 | 330 | 101 | |
XM_005263602.2 | XP_005263659.1 | 1 - 2 | 330 | 101 | |
XM_005263604.2 | XP_005263661.1 | 1 - 2 | 330 | 101 | |
XM_011510667.1 | XP_011508969.1 | 1 - 2 | 330 | 101 | |
XM_011510669.2 | XP_011508971.1 | 1 - 2 | 229 | 101 | |
XM_017003407.1 | XP_016858896.1 | 1 - 2 | 330 | 101 | |
GenBank mRNA | AF452722.1 | - | 1 - 2 | 186 | 101 |
AK225251.1 | - | 1 - 2 | 199 | 101 | |
AK290072.1 | - | 1 - 2 | 330 | 101 | |
AY343910.1 | - | 1 - 2 | 215 | 101 | |
BC120991.1 | - | 1 - 2 | 201 | 101 | |
BC120992.1 | - | 1 - 2 | 201 | 101 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291821.1 | NP_001278750.1 | 1 - 2 | 239 | 101 |
NM_201453.3 | NP_958861.2 | 1 - 2 | 239 | 101 | |
XM_005277637.2 | XP_005277694.1 | 1 - 2 | 318 | 101 | |
XM_005277639.2 | XP_005277696.1 | 1 - 2 | 318 | 101 | |
XM_005277640.2 | XP_005277697.1 | 1 - 2 | 318 | 101 | |
XM_006717116.1 | XP_006717179.1 | 1 - 2 | 318 | 101 | |
XM_017014730.1 | XP_016870219.1 | 1 - 2 | 318 | 101 | |
GenBank mRNA | AK297912.1 | - | 1 - 2 | 201 | 101 |
AY344633.1 | - | 1 - 2 | 215 | 101 | |
BM786305.1 | - | 1 - 2 | 261 | 101 | |
BX509475.1 | - | 1 - 2 | 239 | 101 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024916.3 | NP_001020087.2 | 1 - 2 | 295 | 101 |
NM_001286835.1 | NP_001273764.1 | 1 - 2 | 295 | 101 | |
NM_001286836.1 | NP_001273765.1 | 1 - 2 | 295 | 101 | |
XM_005272748.1 | XP_005272805.1 | 1 - 2 | 255 | 101 | |
XM_005272749.1 | XP_005272806.1 | 1 - 2 | 255 | 101 | |
XM_005272750.1 | XP_005272807.1 | 1 - 2 | 255 | 101 | |
XM_005272751.1 | XP_005272808.1 | 1 - 2 | 255 | 101 | |
XM_006716914.1 | XP_006716977.1 | 1 - 2 | 255 | 101 | |
XM_017014455.1 | XP_016869944.1 | 1 - 2 | 255 | 101 | |
XM_017014458.1 | XP_016869947.1 | 1 - 2 | 255 | 101 | |
XM_017014459.1 | XP_016869948.1 | 1 - 2 | 201 | 101 | |
XM_017014460.1 | XP_016869949.1 | 1 - 2 | 255 | 101 | |
XM_017014461.1 | XP_016869950.1 | 1 - 2 | 201 | 101 | |
GenBank mRNA | AA039734.1 | - | 2 - 3 | 101 | |
AF265442.1 | - | 2 - 3 | 163 | 101 | |
AK292585.1 | - | 1 - 2 | 235 | 101 | |
BC067803.1 | - | 1 - 2 | 233 | 101 | |
BC082271.1 | - | 1 - 2 | 191 | 101 | |
BC141856.1 | - | 1 - 2 | 233 | 101 | |
DA288994.1 | - | 1 - 2 | 255 | 101 | |
DB463072.2 | - | 1 - 2 | 233 | 101 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085457.1 | NP_001078926.1 | 1 - 2 | 258 | 101 |
XM_017015019.1 | XP_016870508.1 | 1 - 2 | 318 | 101 | |
XM_017015020.1 | XP_016870509.1 | 1 - 2 | 318 | 101 | |
XM_017015021.1 | XP_016870510.1 | 1 - 2 | 318 | 101 | |
XM_017015022.1 | XP_016870511.1 | 1 - 2 | 318 | 101 | |
XM_017015023.1 | XP_016870512.1 | 1 - 2 | 318 | 101 | |
XM_017015024.1 | XP_016870513.1 | 1 - 2 | 318 | 101 | |
XM_017015025.1 | XP_016870514.1 | 1 - 2 | 318 | 101 | |
XM_017015026.1 | XP_016870515.1 | 1 - 2 | 318 | 101 | |
XM_017015027.1 | XP_016870516.1 | 1 - 2 | 318 | 101 | |
XM_017015030.1 | XP_016870519.1 | 1 - 2 | 318 | 101 | |
XM_017015032.1 | XP_016870521.1 | 1 - 2 | 318 | 101 | |
XM_017015035.1 | XP_016870524.1 | 1 - 2 | 318 | 101 | |
XM_017015036.1 | XP_016870525.1 | 1 - 2 | 318 | 101 | |
GenBank mRNA | AK293173.1 | - | 1 - 2 | 225 | 101 |
BC069006.1 | - | 1 - 2 | 195 | 101 |