Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- POLR2J2
- Entrez Gene ID
-
246721
- Gene Name
- RNA polymerase II subunit J2
- Gene Aliases
-
HRPB11B, RPB11b1
- Chromosome Location
-
Chr.7: 102636748 - 102671729 on Build GRCh38
- Gene Symbol
- LOC107983993
- Entrez Gene ID
-
107983993
- Gene Name
- uroplakin-3b-like protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 102537919 - 102544881 on Build GRCh38
- Gene Symbol
- POLR2J3
- Entrez Gene ID
-
548644
- Gene Name
- RNA polymerase II subunit J3
- Gene Aliases
-
POLR2J2, RPB11b1, RPB11b2
- Chromosome Location
-
Chr.7: 102537919 - 102572660 on Build GRCh38
- Gene Symbol
- POLR2J4
- Entrez Gene ID
-
84820
- Gene Name
- RNA polymerase II subunit J4, pseudogene
- Gene Aliases
-
RPB11-phi
- Chromosome Location
-
Chr.7: 43940895 - 44019194 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003655.3 |
|
5 - 6 |
647 |
99 |
GenBank mRNA |
AK296066.1 |
- |
4 - 5 |
450 |
99 |
BX537878.1 |
- |
5 - 6 |
602 |
99 |
CD674494.1 |
- |
3 - 4 |
344 |
99 |
- Gene Symbol
- UPK3BL
- Entrez Gene ID
-
100134938
- Gene Name
- uroplakin 3B-like
- Gene Aliases
-
UPLP
- Chromosome Location
-
Chr.7: 102637025 - 102642791 on Build GRCh38
View Full Product Details