Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- KRTAP5-3
- Entrez Gene ID
-
387266
- Gene Name
- keratin associated protein 5-3
- Gene Aliases
-
KRTAP5-9, KRTAP5.3
- Chromosome Location
-
Chr.11: 1607565 - 1608463 on Build GRCh38
- Gene Symbol
- KRTAP5-7
- Entrez Gene ID
-
440050
- Gene Name
- keratin associated protein 5-7
- Gene Aliases
-
KRTAP5-3, KRTAP5.7
- Chromosome Location
-
Chr.11: 71527267 - 71528164 on Build GRCh38
- Gene Symbol
- KRTAP5-8
- Entrez Gene ID
-
57830
- Gene Name
- keratin associated protein 5-8
- Gene Aliases
-
KRTAP5-2, KRTAP5.8, UHSKerB
- Chromosome Location
-
Chr.11: 71538025 - 71539207 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB126077.1 |
- |
1 - 1 |
|
143 |
BC119736.1 |
- |
3 - 3 |
|
143 |
View Full Product Details