Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCE3B
Entrez Gene ID
353143
Gene Name
late cornified envelope 3B
Gene Aliases
LEP14
Chromosome Location
Chr.1: 152613811 - 152614098 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178433.1 NP_848520.1 1 - 1 148 99

View Full Product Details