Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRAMEF18
Entrez Gene ID
391003
Gene Name
PRAME family member 18
Gene Aliases
-
Chromosome Location
Chr.1: 13222695 - 13226154 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099850.2 NP_001093320.2 1 - 2 336 93

Gene Symbol
PRAMEF19
Entrez Gene ID
645414
Gene Name
PRAME family member 19
Gene Aliases
PRAMEF18
Chromosome Location
Chr.1: 13368431 - 13371947 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099790.2 NP_001093260.3 1 - 2 335 93

Gene Symbol
PRAMEF22
Entrez Gene ID
653606
Gene Name
PRAME family member 22
Gene Aliases
PRAMEF3L
Chromosome Location
Chr.1: 13005048 - 13005624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100631.1 NP_001094101.1 1 - 2 294 93

View Full Product Details