Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
TRIM34
Entrez Gene ID
53840
Gene Name
tripartite motif containing 34
Gene Aliases
IFP1, RNF21
Chromosome Location
Chr.11: 5619944 - 5644398 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130390.2 NP_569074.2 4 - 4 853 83
GenBank mRNA AB039904.1 - 4 - 4 835 83

View Full Product Details